accessibility menu, dialog, popup

UserName

NNN

CAS RN 31274-51-8

IUPAC Name: tris({[1,1'-biphenyl]-4-yl})-1,3,5-triazine
Synonyms: tris-biphenyl triazine
Molecular Weight (g/mol): 537.67
Molecular Formula: C39H27N3
InChi Key: CENPSTJGQOQKKW-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results