missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Collidine, 99%

CAS: 108-75-8 | C8H11N | 121.18 g/mol

Supplier:  Thermo Scientific Chemicals A11058AE

 View more versions of this product

Catalog No. AAA11058AE


Only null left
Explore available promotions

Description

Description

2,4,6-Collidine is used as a tissue fixative for electron microscopy. It is useful in dehydrohalogenation reactions and acts as a solvent for the cleavage of hindered esters by anhydrous lithium iodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Collidine is used as a tissue fixative for electron microscopy. It is useful in dehydrohalogenation reactions and acts as a solvent for the cleavage of hindered esters by anhydrous lithium iodide.

Solubility
S
Specifications

Specifications

2,4,6-Collidine
-43°C
9 to 10
54°C (129°F)
C8H11N
MFCD00006338
UN1992
14,9718
Slightly miscible with water. Miscible with ether,methanol,acetone,chloroform,benzene,toluene and dilute acids.
CC1=CC(C)=NC(C)=C1
121.18
121.18
108-75-8
0.916
171°C to 172°C
Unpleasant
1.498
100 mL
107283
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
BWZVCCNYKMEVEX-UHFFFAOYSA-N
2,4,6-trimethylpyridine
7953
99%

Chemical Identifiers

108-75-8
121.18
BWZVCCNYKMEVEX-UHFFFAOYSA-N
7953
CC1=CC(C)=NC(C)=C1
C8H11N
MFCD00006338
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
2,4,6-trimethylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H226-H302-H315-H319-H335;
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H332-H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 203-613-3

RTECSNumber : UU0970000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only