missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Collidine, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 161950025

Catalog No. AC161950025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 4, 6-Collidine
98.5
0.9100g/mL
57°C
98.5% min. (GC)
C8H11N
(C5H2N)(CH3)3
MFCD00006338
01,155; 13,302
15, 9890
Solubility in water: 35g/L (20°C). Other solubilities: soluble in diluted acids,soluble in alcohol,chloroform,benzene and,toluene,miscible with ether
CC1=CC(C)=NC(C)=C1
121.18
121.18
108-75-8
100.0
171.0°C to 172.0°C
Authentic
Glass bottle
1.4995 to 1.5015
2.5L
20, 250
0.91
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
BWZVCCNYKMEVEX-UHFFFAOYSA-N
2,4,6-trimethylpyridine
7953
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 203-613-3

RTECSNumber : UU0970000

TSCA : TSCA

Recommended Storage : This product darkens on standing

SDS
Documents

Documents

Promotions

Promotions