missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Collidine, 99%

Catalog No. p-93088
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC161951000 100 mL
AC161955000 500 mL
AC161950010 1 L
AC161950025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC161951000 Supplier Thermo Scientific Chemicals Supplier No. 161951000
Only null left

CAS: 108-75-8 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-75-8
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00006338
InChI Key BWZVCCNYKMEVEX-UHFFFAOYSA-N
Synonym 2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
PubChem CID 7953
IUPAC Name 2,4,6-trimethylpyridine
SMILES CC1=CC(C)=NC(C)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 171.0°C to 172.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4995 to 1.5015
Linear Formula (C5H2N)(CH3)3
Quantity 100 mL
Beilstein 20, 250
Fieser 01,155; 13,302
Specific Gravity 0.91
Merck Index 15, 9890
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: soluble in diluted acids,soluble in alcohol,chloroform,benzene and,toluene,miscible with ether
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4, 6-Collidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 203-613-3
RTECSNumber UU0970000
TSCA TSCA
Recommended Storage This product darkens on standing

RUO – Research Use Only