missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane 97.0+%, TCI America™

Catalog No. p-7137260
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25235G 5 g
T252325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25235G Supplier TCI America Supplier No. T25235G

Chemical Identifiers

CAS 2554-06-5
Molecular Formula C12H24O4Si4
Molecular Weight (g/mol) 344.66
MDL Number MFCD00040293
InChI Key VMAWODUEPLAHOE-UHFFFAOYSA-N
Synonym 2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetravinyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, cyclotetrasiloxane, 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl, 1,3,5,7-tetravinyl-1,3,5,7-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethyl-1,3,5,7-tetravinylcyclotetrasiloxane, 2,4,6,8-tetravinyl-2,4,6,8-tetramethylcyclotetrasiloxane, tetravinyltetramethylcyclotetrasiloxane, s-tetramethyltetravinylcyclotetrasiloxane, unii-3nt606t1vk, cyclotetrasiloxane, tetraethenyltetramethyl
PubChem CID 75706
IUPAC Name 2,4,6,8-tetrakis(ethenyl)-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
SMILES C[Si]1(O[Si](O[Si](O[Si](O1)(C)C=C)(C)C=C)(C)C=C)C=C

Specifications

Melting Point -44°C
Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 344.66
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane
EINECSNumber (7)-0483
RTECSNumber GZ4550000
TSCA Yes