accessibility menu, dialog, popup

UserName

H3CSiOSiCH3OSiCH3OSiH3COH2CH2CCH2CH2

CAS RN 2554-06-5

IUPAC Name: 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Synonyms: 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular Weight (g/mol): 344.66
Molecular Formula: C12H24O4Si4
InChi Key: VMAWODUEPLAHOE-UHFFFAOYSA-N
SMILES: C[Si]1(O[Si](C)(O[Si](C)(O[Si](C)(O1)C=C)C=C)C=C)C=C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)