missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine 98.0+%, TCI America™

Catalog No. T18105G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18105G Supplier TCI America Supplier No. T18105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32980-71-5
Molecular Formula C6Cl4N4
Molecular Weight (g/mol) 269.894
MDL Number MFCD02093495
InChI Key QNKFHUMDHRWWES-UHFFFAOYSA-N
PubChem CID 97007
IUPAC Name 2,4,6,8-tetrachloropyrimido[5,4-d]pyrimidine
SMILES C12=C(C(=NC(=N1)Cl)Cl)N=C(N=C2Cl)Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 269.89
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6,8-Tetrachloropyrimido[5,4-d]pyrimidine