accessibility menu, dialog, popup

UserName

ClNClNClNClN

CAS RN 32980-71-5

IUPAC Name: tetrachloropyrimido[5,4-d][1,3]diazine
Synonyms: tetrachloropyrimido[5,4-d][1,3]diazine
Molecular Weight (g/mol): 269.89
Molecular Formula: C6Cl4N4
InChi Key: QNKFHUMDHRWWES-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C2N=C(Cl)N=C(Cl)C2=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results