missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triphenylimidazole 98.0+%, TCI America™

$46.60 - $130.48

Chemical Identifiers

CAS 484-47-9
Molecular Formula C21H16N2
Molecular Weight (g/mol) 296.373
MDL Number MFCD00005187
InChI Key RNIPJYFZGXJSDD-UHFFFAOYSA-N
Synonym 2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenyl
PubChem CID 10232
IUPAC Name 2,4,5-triphenyl-1H-imidazole
SMILES C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T09995G
View Documents
TCI America
T09995G
5 g N/A
Only null left
 
T099925G
View Documents
TCI America
T099925G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

484-47-9
296.373
RNIPJYFZGXJSDD-UHFFFAOYSA-N
10232
C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4
C21H16N2
MFCD00005187
2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenyl
2,4,5-triphenyl-1H-imidazole

Specifications

484-47-9
White-Yellow
MFCD00005187
2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenyl
C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4
296.373
296.37
Crystalline Powder
275°C
C21H16N2
5 g
RNIPJYFZGXJSDD-UHFFFAOYSA-N
2,4,5-triphenyl-1H-imidazole
10232
≥98.0% (HPLC,T)
2,4,5-Triphenylimidazole
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NI8710000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'