accessibility menu, dialog, popup

UserName

NHN

CAS RN 484-47-9

IUPAC Name: 2,4,5-triphenyl-1H-imidazole
Synonyms: 1H-imidazole, 2,4,5-triphenyl-
Molecular Weight (g/mol): 296.37
Molecular Formula: C21H16N2
InChi Key: RNIPJYFZGXJSDD-UHFFFAOYSA-N
SMILES: N1C(=NC(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)