Learn More
2,4,5-Triphenylimidazole, 97%
CAS: 484-47-9 | C21H16N2 | 296.373 g/mol
$95.49 - $95.49
Chemical Identifiers
| CAS | 484-47-9 |
|---|---|
| Molecular Formula | C21H16N2 |
| Molecular Weight (g/mol) | 296.373 |
| MDL Number | MFCD00005187 |
| InChI Key | RNIPJYFZGXJSDD-UHFFFAOYSA-N |
| Synonym | 2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenyl |
| PubChem CID | 10232 |
| IUPAC Name | 2,4,5-triphenyl-1H-imidazole |
| SMILES | C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1729109
|
Thermo Scientific Chemicals
A1729109 |
10 g |
Each for $95.49
|
|
|||||
Description
2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Chemical Identifiers
| 484-47-9 | |
| 296.373 | |
| RNIPJYFZGXJSDD-UHFFFAOYSA-N | |
| 10232 | |
| C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4 |
| C21H16N2 | |
| MFCD00005187 | |
| 2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenyl | |
| 2,4,5-triphenyl-1H-imidazole |
Specifications
| 484-47-9 | |
| C21H16N2 | |
| 10 g | |
| 2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenyl | |
| RNIPJYFZGXJSDD-UHFFFAOYSA-N | |
| 2,4,5-triphenyl-1H-imidazole | |
| 10232 | |
| 97% |
| 274°C to 278°C | |
| MFCD00005187 | |
| 236652 | |
| Soluble in methanol. Insoluble in water. | |
| C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4 | |
| 296.373 | |
| 296.38 | |
| 2,4,5-Triphenylimidazole |
Safety and Handling
EINECSNumber : 207-606-6
RTECSNumber : NI8710000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only