missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triphenylimidazole, 97%

Catalog No. AAA1729109
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1729109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1729109 Supplier Thermo Scientific Chemicals Supplier No. A1729109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 484-47-9 | C21H16N2 | 296.373 g/mol

2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

Chemical Identifiers

CAS 484-47-9
Molecular Formula C21H16N2
Molecular Weight (g/mol) 296.373
MDL Number MFCD00005187
InChI Key RNIPJYFZGXJSDD-UHFFFAOYSA-N
Synonym 2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenyl
PubChem CID 10232
IUPAC Name 2,4,5-triphenyl-1H-imidazole
SMILES C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 274°C to 278°C
Quantity 10 g
Beilstein 236652
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 296.38
Percent Purity 97%
Chemical Name or Material 2,4,5-Triphenylimidazole
EINECSNumber 207-606-6
RTECSNumber NI8710000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only