missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethylphenol, 99%

Catalog No. p-7033331
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50427MD 250 mg
AAH5042703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50427MD Supplier Thermo Scientific Chemicals Supplier No. H50427MD
Only null left

CAS: 496-78-6 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 496-78-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00020050
InChI Key VXSCPERJHPWROZ-UHFFFAOYSA-N
Synonym pseudocumenol, phenol, 2,4,5-trimethyl, 5-hydroxypseudocumene, 1-hydroxy-2,4,5-trimethylbenzene, unii-3pca9e425c, acmc-1bmse, 4-06-00-03247 beilstein handbook reference
PubChem CID 10335
IUPAC Name 2,4,5-trimethylphenol
SMILES CC1=CC(=C(C=C1C)O)C

Specifications

Melting Point 72°C
Boiling Point 232°C
Odor Phenol-like
Quantity 250 mg
UN Number UN2430
Formula Weight 136.2
Percent Purity 99%
Chemical Name or Material 2,4,5-Trimethylphenol
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 207-832-5
RTECSNumber SN2815000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only