missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethoxybenzoic acid, 99%

Catalog No. AAA1932409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1932409 10 g
AAA1932418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1932409 Supplier Thermo Scientific Chemicals Supplier No. A1932409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 490-64-2 | C10H11O5 | 211.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 490-64-2
Molecular Formula C10H11O5
Molecular Weight (g/mol) 211.19
MDL Number MFCD00002435
InChI Key KVZUCOGWKYOPID-UHFFFAOYSA-M
Synonym asaronic acid, benzoic acid, 2,4,5-trimethoxy, asarylic acid, calamonic acid, acmc-209ket, 2,4,5-trimethoxybenzoicacid, ksc489o7d, 2,4,5-trimethoxy-benzoic acid, 2,4,5-trimethoxybenzoic acid, alpha-hydroxyphenylacetic acid; amygdalic acid; alpha-hydroxyphenylacetic acid; alpha-hydroxy-benzeneacetic acid
PubChem CID 10276
SMILES COC1=CC(OC)=C(C=C1OC)C([O-])=O

Specifications

Melting Point 143°C to 145°C
Quantity 10 g
IUPAC Name 2,4,5-trimethoxybenzoate
Formula Weight 212.2
Percent Purity 99%
Chemical Name or Material 2,4,5-Trimethoxybenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-715-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only