missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Triiodo-1H-imidazole 98.0+%, TCI America™
Supplier: TCI America T33695G
Specifications
| 2,4,5-Triiodo-1H-imidazole | |
| 186°C | |
| C3HI3N2 | |
| 5 g | |
| HIDCNJIBRZBEPN-UHFFFAOYSA-N | |
| 2,4,5-triiodo-1H-imidazole | |
| 74459 | |
| ≥98.0% (T) |
| 1746-25-4 | |
| White-Yellow | |
| MFCD00068812 | |
| 2811 | |
| C1(=C(N=C(N1)I)I)I | |
| 445.768 | |
| 445.77 | |
| Crystalline Powder |
Chemical Identifiers
| 1746-25-4 | |
| 445.768 | |
| HIDCNJIBRZBEPN-UHFFFAOYSA-N | |
| 2,4,5-triiodo-1H-imidazole |
| C3HI3N2 | |
| MFCD00068812 | |
| 74459 | |
| C1(=C(N=C(N1)I)I)I |
Safety and Handling
RTECSNumber : NI8680000
TSCA : No