missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triiodo-1H-imidazole 98.0+%, TCI America™

Catalog No. p-7137719
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33691G 1 g
T33695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33691G Supplier TCI America Supplier No. T33691G

Chemical Identifiers

CAS 1746-25-4
Molecular Formula C3HI3N2
Molecular Weight (g/mol) 445.768
MDL Number MFCD00068812
InChI Key HIDCNJIBRZBEPN-UHFFFAOYSA-N
PubChem CID 74459
IUPAC Name 2,4,5-triiodo-1H-imidazole
SMILES C1(=C(N=C(N1)I)I)I

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 445.77
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Triiodo-1H-imidazole
RTECSNumber NI8680000
TSCA No