missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trihydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7136949
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17661G 1 g
T17665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17661G Supplier TCI America Supplier No. T17661G
Only null left

Chemical Identifiers

CAS 35094-87-2
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00016592
InChI Key WNCNWLVQSHZVKV-UHFFFAOYSA-N
PubChem CID 643387
IUPAC Name 2,4,5-trihydroxybenzaldehyde
SMILES OC1=CC(O)=C(C=O)C=C1O

Specifications

Color Green-Yellow
Quantity 1 g
Synonym benzaldehyde, 2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxy-benzaldehyde, benzaldehyde,2,4,5-trihydroxy, 2,4,5-trihydroxybenzaldehyde
Formula Weight 154.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trihydroxybenzaldehyde