missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trihydroxybenzaldehyde, 97%

Catalog No. AAA1328306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1328306 5 g
AAA1328303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1328306 Supplier Thermo Scientific Chemicals Supplier No. A1328306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2,4,5-Trihydroxybenzaldehyde, 97%, C7H6O4, CAS Number-35094-87-2, benzaldehyde, 2,4,5-trihydroxy, benzaldehyde,2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxybenzaldehyde, 2,4,5-trihydroxy-benzaldehyde, 5g, 2250084, 154.12, H315-H319-H335 | CAS: 35094-87-2 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35094-87-2
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00016592
InChI Key WNCNWLVQSHZVKV-UHFFFAOYSA-N
PubChem CID 643387
IUPAC Name 2,4,5-trihydroxybenzaldehyde
SMILES OC1=CC(O)=C(C=O)C=C1O

Specifications

Melting Point ∼230°C (decomposition)
Quantity 5 g
Beilstein 2250084
Sensitivity Air Sensitive
Synonym benzaldehyde, 2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxy-benzaldehyde, benzaldehyde,2,4,5-trihydroxy, 2,4,5-trihydroxybenzaldehyde
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2,4,5-Trihydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only