missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorophenylacetic Acid 98.0+%, TCI America™

Catalog No. T24075G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24075G Supplier TCI America Supplier No. T24075G

Chemical Identifiers

CAS 209995-38-0
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00082479
InChI Key YSQLGGQUQDTBSL-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenylacetic acid, 2-2,4,5-trifluorophenyl acetic acid, 2,4,5-trifluorophenyl acetic acid, 2,4,5-trifluoro-phenyl-acetic acid, 2,4,5-trifluorophenylaceticacid, 2,4,5-trifluoro phenyl acetic acid, benzeneacetic acid, 2,4,5-trifluoro, pubchem3526, pubchem7287
PubChem CID 2777950
IUPAC Name 2-(2,4,5-trifluorophenyl)acetic acid
SMILES OC(=O)CC1=CC(F)=C(F)C=C1F

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 5 g
Formula Weight 190.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trifluorophenylacetic Acid