missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorophenylacetic acid, 94%

Catalog No. AAH2619003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2619003 1 g
AAH2619006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2619003 Supplier Thermo Scientific Chemicals Supplier No. H2619003

CAS: 209995-38-0 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 209995-38-0
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00082479
InChI Key YSQLGGQUQDTBSL-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenylacetic acid, 2-2,4,5-trifluorophenyl acetic acid, 2,4,5-trifluorophenyl acetic acid, 2,4,5-trifluoro-phenyl-acetic acid, 2,4,5-trifluorophenylaceticacid, 2,4,5-trifluoro phenyl acetic acid, benzeneacetic acid, 2,4,5-trifluoro, pubchem3526, pubchem7287
PubChem CID 2777950
IUPAC Name 2-(2,4,5-trifluorophenyl)acetic acid
SMILES OC(=O)CC1=CC(F)=C(F)C=C1F

Specifications

Melting Point 121°C to 124°C
Quantity 1 g
Formula Weight 190.12
Percent Purity 94%
Chemical Name or Material 2,4,5-Trifluorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only