missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzyl Chloride 98.0+%, TCI America™

Catalog No. p-7137466
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28891G 1 g
T28895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28891G Supplier TCI America Supplier No. T28891G

Chemical Identifiers

CAS 243139-71-1
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.55
MDL Number MFCD00236319
InChI Key JMXPOOVDUVHJRO-UHFFFAOYSA-N
Synonym 2,4,5-trifluorobenzyl chloride, 1-chloromethyl-2,4,5-trifluorobenzene, 2,4,5-trifluorobenzylchloride, 2,4,5-trifluorobenzyl choride, benzene, 1-chloromethyl-2,4,5-trifluoro, alpha-chloro-2,4,5-trifluorotoluene, 1-chloromethyl-2,4,5-trifluoro-benzene, pubchem4024, acmc-1cjlx
PubChem CID 2777063
IUPAC Name 1-(chloromethyl)-2,4,5-trifluorobenzene
SMILES FC1=CC(F)=C(CCl)C=C1F

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 180.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trifluorobenzyl Chloride