missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzoic acid, 98%

Catalog No. AAA1419306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1419306 5 g
AAA1419314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1419306 Supplier Thermo Scientific Chemicals Supplier No. A1419306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 446-17-3 | C7H2F3O2 | 175.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 446-17-3
Molecular Formula C7H2F3O2
Molecular Weight (g/mol) 175.09
MDL Number MFCD00013306
InChI Key AKAMNXFLKYKFOJ-UHFFFAOYSA-M
Synonym 2,4,5-trifluoro-benzoic acid, benzoic acid, 2,4,5-trifluoro, pubchem1346, acmc-209jyu, 2,4,5-trifluorobenzoicacid, ksc235o3j, 2,4,5-trifluoro benzoic acid, rarechem al bo 0258, akamnxflkykfoj-uhfffaoysa, 2,4,5-trifluorobenzoic acid
PubChem CID 521170
IUPAC Name 2,4,5-trifluorobenzoic acid
SMILES [O-]C(=O)C1=CC(F)=C(F)C=C1F

Specifications

Melting Point 95°C to 100°C
Quantity 5 g
Beilstein 3257609
Formula Weight 176.1
Percent Purity 98%
Chemical Name or Material 2,4,5-Trifluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes

RUO – Research Use Only