missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzeneboronic acid, 97%

CAS: 247564-72-3 | C6H4BF3O2 | 175.90 g/mol

$280.68 - $916.19

Chemical Identifiers

CAS 247564-72-3
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.90
MDL Number MFCD01863165
InChI Key KCHHKNCSISEAAE-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74
PubChem CID 2783133
IUPAC Name (2,4,5-trifluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(F)C=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2785806
View Documents
Thermo Scientific Chemicals
H2785806
5 g N/A
Only null left
 
AAH2785814
View Documents
Thermo Scientific Chemicals
H2785814
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

247564-72-3
175.90
KCHHKNCSISEAAE-UHFFFAOYSA-N
2783133
OB(O)C1=CC(F)=C(F)C=C1F
C6H4BF3O2
MFCD01863165
2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74
(2,4,5-trifluorophenyl)boronic acid

Specifications

247564-72-3
C6H4BF3O2
5 g
KCHHKNCSISEAAE-UHFFFAOYSA-N
(2,4,5-trifluorophenyl)boronic acid
2783133
97%
99°C to 101°C
MFCD01863165
2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74
OB(O)C1=CC(F)=C(F)C=C1F
175.90
175.9
2,4,5-Trifluorobenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only