missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzeneboronic acid, 97%

Catalog No. AAH2785814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2785814 25 g
AAH2785806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2785814 Supplier Thermo Scientific Chemicals Supplier No. H2785814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 247564-72-3 | C6H4BF3O2 | 175.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 247564-72-3
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.90
MDL Number MFCD01863165
InChI Key KCHHKNCSISEAAE-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74
PubChem CID 2783133
IUPAC Name (2,4,5-trifluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(F)C=C1F

Specifications

Melting Point 99°C to 101°C
Quantity 25 g
Formula Weight 175.9
Percent Purity 97%
Chemical Name or Material 2,4,5-Trifluorobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only