missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluoroaniline 98.0+%, TCI America™

Catalog No. T16455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16455G Supplier TCI America Supplier No. T16455G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 367-34-0
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00007649
InChI Key QMYVWJVVVMIBMM-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenylamine, benzenamine, 2,4,5-trifluoro, pubchem2469, 2,4,5-triflouroaniline, 2,4,5-trifloro aniline, acmc-1cs4m, 2,4,5-trifluoro-aniline, 2,4,5-trifluoro aniline, ksc497k9p, 2,4,5-trifluoro-phenylamine
PubChem CID 94953
IUPAC Name 2,4,5-trifluoroaniline
SMILES C1=C(C(=CC(=C1F)F)F)N

Specifications

Melting Point 60°C
Quantity 5 g
Formula Weight 147.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trifluoroaniline