missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Trifluoro-3-hydroxybenzoic Acid 98.0+%, TCI America™
Supplier: TCI America T238025G
Specifications
| 2,4,5-Trifluoro-3-hydroxybenzoic Acid | |
| 146°C | |
| C7H3F3O3 | |
| 25 g | |
| YYAFUGSJSHXYNK-UHFFFAOYSA-N | |
| 2,4,5-trifluoro-3-hydroxybenzoic acid | |
| 2734270 | |
| ≥98.0% (GC,T) |
| 116751-24-7 | |
| White-Yellow | |
| MFCD00800378 | |
| 3-hydroxy-2,4,5-trifluorobenzoic acid, 3-hydroxy-2,4,5-trifluorobenzoicacid, benzoic acid, 2,4,5-trifluoro-3-hydroxy, pubchem1353, 2,4,5-trifluoro-3-hydroxy-benzoic acid, acmc-2099tc, ksc496g3r, 2.4,5-trifluoro-3-hydroxybenzoic acid | |
| C1=C(C(=C(C(=C1F)F)O)F)C(=O)O | |
| 192.093 | |
| 192.09 | |
| Crystalline Powder |
Chemical Identifiers
| 116751-24-7 | |
| 192.093 | |
| YYAFUGSJSHXYNK-UHFFFAOYSA-N | |
| 2734270 | |
| C1=C(C(=C(C(=C1F)F)O)F)C(=O)O |
| C7H3F3O3 | |
| MFCD00800378 | |
| 3-hydroxy-2,4,5-trifluorobenzoic acid, 3-hydroxy-2,4,5-trifluorobenzoicacid, benzoic acid, 2,4,5-trifluoro-3-hydroxy, pubchem1353, 2,4,5-trifluoro-3-hydroxy-benzoic acid, acmc-2099tc, ksc496g3r, 2.4,5-trifluoro-3-hydroxybenzoic acid | |
| 2,4,5-trifluoro-3-hydroxybenzoic acid |
Safety and Handling
EINECSNumber : (3)-4469
TSCA : No