missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluoro-3-hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. T23805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23805G 5 g
T238025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23805G Supplier TCI America Supplier No. T23805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116751-24-7
Molecular Formula C7H3F3O3
Molecular Weight (g/mol) 192.093
MDL Number MFCD00800378
InChI Key YYAFUGSJSHXYNK-UHFFFAOYSA-N
Synonym 3-hydroxy-2,4,5-trifluorobenzoic acid, 3-hydroxy-2,4,5-trifluorobenzoicacid, benzoic acid, 2,4,5-trifluoro-3-hydroxy, pubchem1353, 2,4,5-trifluoro-3-hydroxy-benzoic acid, acmc-2099tc, ksc496g3r, 2.4,5-trifluoro-3-hydroxybenzoic acid
PubChem CID 2734270
IUPAC Name 2,4,5-trifluoro-3-hydroxybenzoic acid
SMILES C1=C(C(=C(C(=C1F)F)O)F)C(=O)O

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 5 g
Formula Weight 192.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trifluoro-3-hydroxybenzoic Acid
EINECSNumber (3)-4469
TSCA No