missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorothiophenol, 97%

Catalog No. aal0332403
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
AAL0332403 1 g
AAL0332406 5 g
2 options

Catalog No. AAL0332403

Supplier: Thermo Scientific Chemicals L0332403

Only null left

CAS: 3773-14-6 | C6H3Cl3S | 213.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3773-14-6
Molecular Formula C6H3Cl3S
Molecular Weight (g/mol) 213.5
MDL Number MFCD00014447
InChI Key JARIALSGFXECCH-UHFFFAOYSA-N
Synonym 2,4,5-trichlorothiophenol, renacit ii, benzenethiol, 2,4,5-trichloro, 2,4,5-trichlorobenzene-1-thiol, acmc-209iv9, 2,4,5-trichloro-thiophenol, 2,4,5-trichlorophenyl mercaptan, 2,4,5-tris chloranyl benzenethiol, n-memet-n-trimetsilylmet benzylamine
PubChem CID 19597
IUPAC Name 2,4,5-trichlorobenzenethiol
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S

Specifications

Melting Point 112°C to 118°C
Boiling Point 153°C to 154°C (15 mmHg)
Odor Unpleasant
Quantity 1 g
UN Number UN3335
Beilstein 509727
Sensitivity Air Sensitive
Formula Weight 213.51
Percent Purity 97%
Chemical Name or Material 2,4,5-Trichlorothiophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
EINECSNumber 223-223-7
RTECSNumber DC1948000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only