missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichloropyrimidine 98.0+%, TCI America™

Catalog No. p-7137253
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25125G 5 g
T251225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25125G Supplier TCI America Supplier No. T25125G
Only null left

Chemical Identifiers

CAS 5750-76-5
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.416
MDL Number MFCD03788200
InChI Key GIKMWFAAEIACRF-UHFFFAOYSA-N
Synonym 2,5,6-trichloropyrimidine, 2,4,5-trichloro-pyrimidine, trichloropyrimidine, pyrimidine, 2,4,5-trichloro, 2,4,5-trichlorpyrimidin, pubchem5302, zlchem 1349, pyrimidine, trichloro, acmc-209lys, 2,5-trichloropyrimidine
PubChem CID 237259
IUPAC Name 2,4,5-trichloropyrimidine
SMILES C1=C(C(=NC(=N1)Cl)Cl)Cl

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 g
Formula Weight 183.42
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trichloropyrimidine