accessibility menu, dialog, popup

UserName

CAS RN 5750-76-5

IUPAC Name: 2,4,5-trichloropyrimidine
Synonyms: 2,4,5-trichloropyrimidine
Molecular Weight (g/mol): 183.42
Molecular Formula: C4HCl3N2
InChi Key: GIKMWFAAEIACRF-UHFFFAOYSA-N
SMILES: ClC1=NC=C(Cl)C(Cl)=N1
Boiling Point 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)