missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorophenol 95.0+%, TCI America™

Catalog No. p-7136254
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
T038925G 25 g
T0389500G 500 g
2 options

Catalog No. T038925G

Supplier: TCI America T038925G

Only null left

Chemical Identifiers

CAS 95-95-4
Molecular Formula C6H3Cl3O
Molecular Weight (g/mol) 197.44
MDL Number MFCD00002170
InChI Key LHJGJYXLEPZJPM-UHFFFAOYSA-N
Synonym dowicide 2, collunosol, preventol i, phenol, 2,4,5-trichloro, 2,4,5-trichloro phenol, rcra waste number u230, 2,4,5-trichloro-phenol, ccris 718, rcra waste no. u230, dsstox_cid_4359
PubChem CID 7271
ChEBI CHEBI:28520
IUPAC Name 2,4,5-trichlorophenol
SMILES OC1=CC(Cl)=C(Cl)C=C1Cl

Specifications

Melting Point 67°C
Color White
Boiling Point 115°C
Quantity 25 g
UN Number 2020
Formula Weight 197.44
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trichlorophenol
EINECSNumber (3)-0931
RTECSNumber SN1400000
TSCA Yes