missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorobenzenethiol 97.0+%, TCI America™

Catalog No. T072825G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T072825G 25 g
T072810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T072825G Supplier TCI America Supplier No. T072825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3773-14-6
Molecular Formula C6H3Cl3S
Molecular Weight (g/mol) 213.5
MDL Number MFCD00014447
InChI Key JARIALSGFXECCH-UHFFFAOYSA-N
Synonym 2,4,5-trichlorothiophenol, renacit ii, benzenethiol, 2,4,5-trichloro, 2,4,5-trichlorobenzene-1-thiol, acmc-209iv9, 2,4,5-trichloro-thiophenol, 2,4,5-trichlorophenyl mercaptan, 2,4,5-tris chloranyl benzenethiol, n-memet-n-trimetsilylmet benzylamine
PubChem CID 19597
IUPAC Name 2,4,5-trichlorobenzenethiol
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S

Specifications

Melting Point 114°C
Color White-Yellow
Boiling Point 154°C
Quantity 25 g
Formula Weight 213.50
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trichlorobenzenethiol
RTECSNumber DC1948000
TSCA No