missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorobenzenesulfonyl chloride, 98%

Catalog No. p-7024544
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1939006 5 g
AAA1939022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1939006 Supplier Thermo Scientific Chemicals Supplier No. A1939006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15945-07-0 | C6H2Cl4O2S | 279.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15945-07-0
Molecular Formula C6H2Cl4O2S
Molecular Weight (g/mol) 279.94
MDL Number MFCD00007428
InChI Key WNVVRCKTQSCPAC-UHFFFAOYSA-N
Synonym 2,4,5-trichlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,5-trichloro, unii-7gd5283xix, 2,4,5-trichlorobenzenesulphonyl chloride, 2,4,5-trichlorobenzenesulfonylchloride, pubchem5174, acmc-1c50t, 2,4,5trichlorophenylsulfonylchloride, 2,5-trichlorobenzenesulfonyl chloride, chloro 2,4,5-trichlorophenyl sulfone
PubChem CID 27575
IUPAC Name 2,4,5-trichlorobenzenesulfonyl chloride
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)Cl

Specifications

Melting Point 67°C to 71°C
Quantity 5 g
UN Number UN3261
Beilstein 1112595
Sensitivity Moisture sensitive
Formula Weight 279.96
Percent Purity 98%
Chemical Name or Material 2,4,5-Trichlorobenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 240-079-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only