missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%

Catalog No. p-195508
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC137930050 5 g Glass bottle
AC137930250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC137930050 Supplier Thermo Scientific Chemicals Supplier No. 137930050
Only null left

CAS: 5392-28-9 | C4H10N6O4S | 238.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5392-28-9
Molecular Formula C4H10N6O4S
Molecular Weight (g/mol) 238.22
MDL Number MFCD06408008
InChI Key MQEFDQWUCTUJCP-UHFFFAOYSA-N
Synonym 2,4,5,6-tetraaminopyrimidine sulfate, pyrimidinetetramine sulfate, pyrimidine-2,4,5,6-tetraamine sulfate, pyrimidine-2,4,5,6-tetramine; sulfuric acid, tetraaminopyrimidine sulfate, pyrimidinetetramine, sulfate 1:1, pyrimidinetetramine, sulfate, tetraaminopyrimidine sulphate, unii-53qqy0i99m, tetra-aminopyrimidine sulfate
PubChem CID 79358
SMILES OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Ochre to Brown
Loss on Drying 9% max. (1 g, 150°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 25, 423
Solubility Information Solubility in water: slightly soluble
IUPAC Name pyrimidine-2,4,5,6-tetramine;sulfuric acid
Formula Weight 238.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 226-393-0
TSCA TSCA

RUO – Research Use Only