missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 137930050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC137930050


Only null left

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5392-28-9
238.22
MQEFDQWUCTUJCP-UHFFFAOYSA-N
79358
OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1
C4H10N6O4S
MFCD00012786
2,4,5,6-tetraaminopyrimidine sulfate, pyrimidinetetramine sulfate, pyrimidine-2,4,5,6-tetraamine sulfate, pyrimidine-2,4,5,6-tetramine; sulfuric acid, tetraaminopyrimidine sulfate, pyrimidinetetramine, sulfate 1:1, pyrimidinetetramine, sulfate, tetraaminopyrimidine sulphate, unii-53qqy0i99m, tetra-aminopyrimidine sulfate
pyrimidine-2,4,5,6-tetramine;sulfuric acid

Specifications

2,4,5,6-Tetraaminopyrimidine sulfate hydrate
5392-28-9
100.0
Authentic
Glass bottle
MFCD00012786
25, 423
Solubility in water: slightly soluble
OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1
238.22
238.22
98%
97.5
9% max. (1 g, 150°C, 3 hrs)
98%
C4H10N6O4S
5g
2,4,5,6-tetraaminopyrimidine sulfate, pyrimidinetetramine sulfate, pyrimidine-2,4,5,6-tetraamine sulfate, pyrimidine-2,4,5,6-tetramine; sulfuric acid, tetraaminopyrimidine sulfate, pyrimidinetetramine, sulfate 1:1, pyrimidinetetramine, sulfate, tetraaminopyrimidine sulphate, unii-53qqy0i99m, tetra-aminopyrimidine sulfate
MQEFDQWUCTUJCP-UHFFFAOYSA-N
pyrimidine-2,4,5,6-tetramine;sulfuric acid
79358
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-393-0

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions