missing translation for 'onlineSavingsMsg'
Learn More

2,4,5,6-Tetraaminopyrimidine sulfate, 97%

Catalog No. p-7024334
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1878809 10 g
AAA1878818 50 g
AAA1878830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1878809 Supplier Thermo Scientific Chemicals Supplier No. A1878809

CAS: 5392-28-9 | C4H10N6O4S | 238.22 g/mol

Used as an intermediate in the production of Methotrexate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an intermediate in the production of Methotrexate.

Solubility
Soluble in water and dimethyl sulfoxide.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 5392-28-9
Molecular Formula C4H10N6O4S
Molecular Weight (g/mol) 238.22
MDL Number MFCD06408008
InChI Key MQEFDQWUCTUJCP-UHFFFAOYSA-N
Synonym 2,4,5,6-tetraaminopyrimidine sulfate, pyrimidinetetramine sulfate, pyrimidine-2,4,5,6-tetraamine sulfate, pyrimidine-2,4,5,6-tetramine; sulfuric acid, tetraaminopyrimidine sulfate, pyrimidinetetramine, sulfate 1:1, pyrimidinetetramine, sulfate, tetraaminopyrimidine sulphate, unii-53qqy0i99m, tetra-aminopyrimidine sulfate
PubChem CID 79358
IUPAC Name pyrimidine-2,4,5,6-tetramine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1

Specifications

Melting Point >300°C
Quantity 10 g
Beilstein 3785189
Solubility Information Soluble in water and dimethyl sulfoxide.
Formula Weight 238.23
Percent Purity 97%
Chemical Name or Material 2,4,5,6-Tetraaminopyrimidine sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-393-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only