Learn More
2,4,4-Trimethylcyclopentanone, 98%
CAS: 4694-12-6 | C8H14O | 126.20 g/mol
$75.03 - $282.45
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2042503
|
Thermo Scientific Chemicals
B2042503 |
1 g |
Each for $75.03
|
|
|||||
|
AAB2042506
|
Thermo Scientific Chemicals
B2042506 |
5 g |
Each for $282.45
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4694-12-6 | |
| MFCD00019313 |
Specifications
| 4694-12-6 | |
| 159°C to 161°C | |
| C7H16O2 | |
| MFCD00019313 | |
| UN1224 | |
| MBNMGGKBGCIEGF-UHFFFAOYSA-N | |
| 1,1-diethoxypropane | |
| 20858 | |
| 98% |
| 0.877 | |
| 43°C (110°F) | |
| 1.432 | |
| 1 g | |
| propionaldehyde diethyl acetal, propane, 1,1-diethoxy, propanaldiethylacetal, propionaldehyde, diethyl acetal, diethyl propional, propanal diethyl acetal, propionaldehyde diethylacetal, propane,1-diethoxy, propioaldehyde diethylacetal, propioaldehyde diethyl acetal | |
| CCC(OCC)OCC | |
| 132.20 | |
| 126.2 | |
| 2,4,4-Trimethylcyclopentanone |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only