accessibility menu, dialog, popup

UserName

CAS RN 4694-12-6

H3CH3CCH3O

CAS RN 4694-12-6

IUPAC Name: 2,4,4-trimethylcyclopentan-1-one
Synonyms: 2,4,4-trimethylcyclopentan-1-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: OXTQEWUBDTVSFB-UHFFFAOYNA-N
SMILES: CC1CC(C)(C)CC1=O
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)