missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethylcyclopentanone, 98%

Catalog No. AAB2042506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2042506 5 g
AAB2042503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2042506 Supplier Thermo Scientific Chemicals Supplier No. B2042506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4694-12-6 | C8H14O | 126.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4694-12-6
MDL Number MFCD00019313

Specifications

Density 0.877
Boiling Point 159°C to 161°C
Flash Point 43°C (110°F)
Molecular Formula C8H14O
Refractive Index 1.432
Quantity 5 g
UN Number UN1224
Synonym 2,4,4-trimethylcyclopentanone, cyclopentanone, 2,4,4-trimethyl, acmc-1aq0h, 1.1.3-trimethylcyclopentanon-4, cyclopentanone,2,4,4-trimethyl, 2,4,4-trimethyl-cyclopentan-1-one, 2,4,4-trimethyl cyclopentanone
InChI Key OXTQEWUBDTVSFB-UHFFFAOYNA-N
SMILES CC1CC(CC1=O)(C)C
IUPAC Name 2,4,4-trimethylcyclopentan-1-one
Molecular Weight (g/mol) 126.20
PubChem CID 107324
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material 2,4,4-Trimethylcyclopentanone
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only