missing translation for 'onlineSavingsMsg'
Learn More

2,4,4'-Trichloro-2'-hydroxydiphenyl Ether 98.0+%, TCI America™

Catalog No. p-7136994
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T187225G 25 g
T1872100G 100 g
T1872500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T187225G Supplier TCI America Supplier No. T187225G
Only null left

Chemical Identifiers

CAS 3380-34-5
Molecular Formula C12H7Cl3O2
Molecular Weight (g/mol) 289.536
MDL Number MFCD00800992
InChI Key XEFQLINVKFYRCS-UHFFFAOYSA-N
Synonym triclosan, 5-chloro-2-2,4-dichlorophenoxy phenol, irgasan, 2,4,4'-trichloro-2'-hydroxydiphenyl ether, cloxifenolum, triclosanum, irgasan dp300, stri-dex face wash, aquasept, sapoderm
PubChem CID 5564
ChEBI CHEBI:164200
IUPAC Name 5-chloro-2-(2,4-dichlorophenoxy)phenol
SMILES C1=CC(=C(C=C1Cl)O)OC2=C(C=C(C=C2)Cl)Cl

Specifications

Melting Point 57°C
Color White
Boiling Point 290°C
Quantity 25 g
UN Number 3077
Formula Weight 289.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,4′-Trichloro-2′-hydroxydiphenyl Ether
EINECSNumber (9)-0381&(9)-0922
RTECSNumber KO1100000
TSCA Yes