missing translation for 'onlineSavingsMsg'
Learn More

2,4,4,6-Tetrabromo-2,5-cyclohexadienone 97.0+%, TCI America™

Catalog No. T12355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12355G 5 g
T123525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12355G Supplier TCI America Supplier No. T12355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20244-61-5
Molecular Formula C6H2Br4O
Molecular Weight (g/mol) 409.697
MDL Number MFCD00001589
InChI Key NJQJGRGGIUNVAB-UHFFFAOYSA-N
Synonym 2,4,4,6-tetrabromocyclohexa-2,5-dienone, 2,4,4,6-tetrabromo-2,5-cyclohexadienone, 2,4,4,6-tetrabromo-2,5-cyclohexadien-1-one, 2,5-cyclohexadien-1-one, 2,4,4,6-tetrabromo, 2,2,4,4-tetrabromo-2,5-cyclohexadienone, tbco, acmc-1cce4, ksc496q7n, 2,4,4,6-tetrabromocyclohexadienone, 2,4,4,6-tetrabromocyclohexadiene-1-one
PubChem CID 88433
IUPAC Name 2,4,4,6-tetrabromocyclohexa-2,5-dien-1-one
SMILES C1=C(C(=O)C(=CC1(Br)Br)Br)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 409.70
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,4,6-Tetrabromo-2,5-cyclohexadienone
TSCA No
Recommended Storage Freezer