accessibility menu, dialog, popup

UserName

CAS RN 20244-61-5

IUPAC Name: 2,4,4,6-tetrabromocyclohexa-2,5-dien-1-one
Synonyms: 2,4,4,6-tetrabromocyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 409.7
Molecular Formula: C6H2Br4O
InChi Key: NJQJGRGGIUNVAB-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)(Br)C=C(Br)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results