missing translation for 'onlineSavingsMsg'
Learn More

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97.0+%, TCI America™

Catalog No. p-7137037
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19521G 1 g
T19525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19521G Supplier TCI America Supplier No. T19521G
Only null left

Chemical Identifiers

CAS 269409-97-4
Molecular Formula C12H17BO3
Molecular Weight (g/mol) 220.075
MDL Number MFCD02093754
InChI Key VLROJECCXBBKPZ-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-hydroxyphenylboronic acid pinacol ester, phenol, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2yl phenol, 2-hydroxyphenylboronic acid, pinacol ester, 2-2-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-hydroxybenzeneboronic acid, pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, pubchem23514
PubChem CID 2734622
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2O

Specifications

Color Yellow
Boiling Point 94°C
Quantity 1 g
Formula Weight 220.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol