missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Bromide 98.0+%, TCI America™
Supplier: TCI America T33931G
Specifications
| 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Bromide | |
| 86°C | |
| C13H18BBrO2 | |
| 1 g | |
| ROIXSNLOYHDYBP-UHFFFAOYSA-N | |
| 2-[2-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 3834834 | |
| ≥98.0% (GC,T) |
| 377780-72-8 | |
| White | |
| MFCD01632210 | |
| 2-2-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromomethylphenylboronic acid pinacol ester, 2-bromomethyl benzeneboronic acid, pinacol ester, 2-bromomethylphenylboronic acid, pinacol ester, 2-bromomethyl benzeneboronic acid pinacol ester, 2-bromomethylphenyl boronic acid, pinacol ester, amtb373, 2-bromomethyl phenylboronic pinacol ester, 2-bromoethylphenylboronic acid pinacol ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CBr | |
| 296.999 | |
| 297.00 | |
| Crystalline Powder |
Chemical Identifiers
| 377780-72-8 | |
| 296.999 | |
| ROIXSNLOYHDYBP-UHFFFAOYSA-N | |
| 3834834 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CBr |
| C13H18BBrO2 | |
| MFCD01632210 | |
| 2-2-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromomethylphenylboronic acid pinacol ester, 2-bromomethyl benzeneboronic acid, pinacol ester, 2-bromomethylphenylboronic acid, pinacol ester, 2-bromomethyl benzeneboronic acid pinacol ester, 2-bromomethylphenyl boronic acid, pinacol ester, amtb373, 2-bromomethyl phenylboronic pinacol ester, 2-bromoethylphenylboronic acid pinacol ester | |
| 2-[2-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No