missing translation for 'onlineSavingsMsg'
Learn More

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Bromide 98.0+%, TCI America™

Catalog No. T33935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33935G 5 g
T33931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33935G Supplier TCI America Supplier No. T33935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 377780-72-8
Molecular Formula C13H18BBrO2
Molecular Weight (g/mol) 296.999
MDL Number MFCD01632210
InChI Key ROIXSNLOYHDYBP-UHFFFAOYSA-N
Synonym 2-2-bromomethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromomethylphenylboronic acid pinacol ester, 2-bromomethyl benzeneboronic acid, pinacol ester, 2-bromomethylphenylboronic acid, pinacol ester, 2-bromomethyl benzeneboronic acid pinacol ester, 2-bromomethylphenyl boronic acid, pinacol ester, amtb373, 2-bromomethyl phenylboronic pinacol ester, 2-bromoethylphenylboronic acid pinacol ester
PubChem CID 3834834
IUPAC Name 2-[2-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2CBr

Specifications

Melting Point 86°C
Color White
Quantity 5 g
Formula Weight 297.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl Bromide