missing translation for 'onlineSavingsMsg'
Learn More

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 97.0+%, TCI America™

Catalog No. T31611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31611G 1 g
T31615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31611G Supplier TCI America Supplier No. T31611G

Chemical Identifiers

CAS 380151-85-9
Molecular Formula C13H17BO3
Molecular Weight (g/mol) 232.09
MDL Number MFCD07363841
InChI Key SLJMPQLPRHIAOM-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-formylphenylboronic acid pinacol ester, 2-formylphenyl boronic acid pinacol ester, 2-formylphenylboronic acid, pinacol ester, 2-formylbenzeneboronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-2-formylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzaldehyde, amtb063, 2-boronobenzaldehyde, pinacol ester
PubChem CID 10977322
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1C=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 232.09
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
TSCA No
Recommended Storage Refrigerator