missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
$42.87 - $124.89
Chemical Identifiers
| CAS | 94242-85-0 |
|---|---|
| Molecular Formula | C7H15BO2 |
| Molecular Weight (g/mol) | 142.005 |
| MDL Number | MFCD09027082 |
| InChI Key | FOQJHZPURACERJ-UHFFFAOYSA-N |
| PubChem CID | 11759390 |
| IUPAC Name | 2,4,4,5,5-pentamethyl-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C |
Chemical Identifiers
| 94242-85-0 | |
| 142.005 | |
| FOQJHZPURACERJ-UHFFFAOYSA-N | |
| 2,4,4,5,5-pentamethyl-1,3,2-dioxaborolane |
| C7H15BO2 | |
| MFCD09027082 | |
| 11759390 | |
| B1(OC(C(O1)(C)C)(C)C)C |
Specifications
| 94242-85-0 | |
| 35°C | |
| MFCD09027082 | |
| 3272 | |
| B1(OC(C(O1)(C)C)(C)C)C | |
| 142.005 | |
| 142.01 | |
| Liquid |
| Colorless | |
| C7H15BO2 | |
| 1 g | |
| FOQJHZPURACERJ-UHFFFAOYSA-N | |
| 2,4,4,5,5-pentamethyl-1,3,2-dioxaborolane | |
| 11759390 | |
| ≥98.0% (GC) | |
| 2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator