missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 35°C |
| Quantity | 1 g |
| UN Number | 3272 |
| Formula Weight | 142.01 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,4,4,5,5-Pentamethyl-1,3,2-dioxaborolane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |