missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethanesulfonic Acid 99.0+%, TCI America™

Catalog No. H039625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H039625G 25 g
H0396500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H039625G Supplier TCI America Supplier No. H039625G

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Melting Point 210°C
Color White
Quantity 25 g
Formula Weight 238.30
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethanesulfonic Acid [Good′s buffer component for biologica
RTECSNumber TL6809000
TSCA Yes