missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Thienyl)ethanol, 97%

Catalog No. AC211710050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211710050 5 g Glass bottle
AC211710250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC211710050 Supplier Thermo Scientific Chemicals Supplier No. 211710050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13781-67-4 | C6H8OS | 128.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13781-67-4
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00009766
InChI Key YYPNNBPPDFTQFX-UHFFFAOYSA-N
Synonym 2-thiophen-3-yl ethanol, 3-thiopheneethanol, 2-3-thienyl ethanol, thiophene-3-ethanol, 3-2-hydroxyethyl thiophene, 2-thiophen-3-yl ethan-1-ol, 2-thien-3-yl ethan-1-ol, 3-thiophene-ethanol, pubchem20415
PubChem CID 83731
SMILES OCCC1=CSC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 1.1400g/mL
Boiling Point 110.0°C to 111.0°C (14.0 mmHg)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5510 to 1.5530
Quantity 5 g
Beilstein 17, IV, 1277
Specific Gravity 1.14
IUPAC Name 2-(thiophen-3-yl)ethan-1-ol
Formula Weight 128.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(3-Thienyl)ethanol
Show More Show Less
EINECSNumber 237-434-7

RUO – Research Use Only